siRNA Details
| Gene Symbol: GINS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126129.1 | NP_001119601.1 | 3 | 528 | ||
| NM_022770.3 | NP_073607.2 | 2 | 411 | |||
| GenBank mRNA | AK127454.1 | - | 1 | 173 | ||
| AK315622.1 | - | 2 | 362 | |||
| BC005879.1 | - | 2 | 286 | 3529058 | ||
| BC014437.1 | - | 2 | 305 | 4870865 | ||
| BP374281.1 | - | 3 | 471 | |||
| DA550725.1 | - | 2 | 411 | |||
| KU178752.1 | - | 2 | 203 | |||
| KU178753.1 | - | 3 | 320 | |||
| KU178754.1 | - | 3 | 262 | |||

GINS3
Genomic Map