siRNA Details
| Gene Symbol: THOC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142350.1 | NP_001135822.1 | 4 | 558 | ||
| NM_024339.3 | NP_077315.2 | 4 | 558 | |||
| GenBank mRNA | AK075330.1 | - | 4 | 525 | ||
| AK308960.1 | - | 2 | 720 | |||
| AK310113.1 | - | 2 | 732 | |||
| AK314086.1 | - | 4 | 536 | |||
| AL832548.1 | - | 3 | 162 | |||
| BC003118.1 | - | 5 | 438 | 3537243 | ||
| BC050674.1 | - | 4 | 497 | 6067702 | ||
| BG613317.1 | - | 4 | 563 | |||

THOC6
Genomic Map