siRNA Details
| Gene Symbol: ARMC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304271.1 | NP_001291200.1 | 2 | 488 | ||
| NM_001304272.1 | NP_001291201.1 | 2 | 488 | |||
| NM_001304273.1 | NP_001291202.1 | 2 | 488 | |||
| NM_024585.3 | NP_078861.1 | 2 | 488 | |||
| GenBank mRNA | AK025813.1 | - | 2 | 296 | ||
| AK300997.1 | - | 2 | 488 | |||
| BC011728.1 | - | 2 | 453 | 4423125 | ||
| BI826938.1 | - | 2 | 454 | |||

ARMC7
Genomic Map