siRNA Details
| Gene Symbol: ZNF750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024702.2 | NP_078978.2 | 2, 3 | 1755 | ||
| GenBank mRNA | AK023903.1 | - | 2, 3 | 1724 | ||
| AK301690.1 | - | 2, 3 | 553 | |||
| BC109036.1 | - | 2, 3 | 1714 | 40028398 | ||
| BC109037.1 | - | 2, 3 | 1714 | 40028404 | ||

ZNF750
Genomic Map