siRNA Details
| Gene Symbol: C12orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024738.2 | NP_079014.1 | 5 | 2767 | ||
| XM_005253937.3 | XP_005253994.1 | 4 | 2604 | |||
| GenBank mRNA | AK025068.1 | - | 5 | 2735 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C12orf49| Gene Symbol: C12orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024738.2 | NP_079014.1 | 5 | 2767 | ||
| XM_005253937.3 | XP_005253994.1 | 4 | 2604 | |||
| GenBank mRNA | AK025068.1 | - | 5 | 2735 | ||