siRNA Details
| Gene Symbol: SHCBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324319.1 | NP_001311248.1 | 2 | 205 | ||
| NM_024745.4 | NP_079021.3 | 2 | 202 | |||
| NR_136738.1 | 2 | 205 | ||||
| XM_011523335.2 | XP_011521637.1 | 2 | 3736 | |||
| GenBank mRNA | AK055931.1 | - | 2 | 173 | ||
| AK292164.1 | - | 2 | 169 | |||
| AK310430.1 | - | 2 | 171 | |||
| AK315706.1 | - | 2 | 171 | |||
| BC030699.1 | - | 2 | 177 | 4829528 | ||
| BP245197.1 | - | 2 | 202 | |||

SHCBP1
Genomic Map