siRNA Details
| Gene Symbol: KDM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145348.1 | NP_001138820.1 | 3 | 746 | ||
| NM_024773.2 | NP_079049.2 | 3 | 696 | |||
| XM_017023676.1 | XP_016879165.1 | 3 | 696 | |||
| XM_017023677.1 | XP_016879166.1 | 4 | 781 | |||
| XM_017023678.1 | XP_016879167.1 | 4 | 777 | |||
| XM_017023679.1 | XP_016879168.1 | 4 | 777 | |||
| GenBank mRNA | AK023860.1 | - | 3 | 687 | ||
| AK225066.1 | - | 3 | 653 | |||
| AK297166.1 | - | 3 | 746 | |||
| BC027911.1 | - | 3 | 675 | 5207043 | ||
| KU178785.1 | - | 2 | 514 | |||
| KU178786.1 | - | 2 | 514 | |||

KDM8
Genomic Map