siRNA Details
| Gene Symbol: TCTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143850.2 | NP_001137322.1 | 3 | 345 | ||
| NM_024809.4 | NP_079085.2 | 3 | 345 | |||
| XM_005253623.2 | XP_005253680.1 | 3 | 345 | |||
| XM_006719605.3 | XP_006719668.1 | 3 | 345 | |||
| XM_017019974.1 | XP_016875463.1 | 3 | 345 | |||
| XM_017019975.1 | XP_016875464.1 | 3 | 350 | |||
| GenBank mRNA | AK056924.1 | - | 3 | 343 | ||
| AK292153.1 | - | 3 | 345 | |||
| BC009112.1 | - | 3 | 314 | 3463475 | ||
| DQ278870.1 | - | 3 | 343 | |||

TCTN2
Genomic Map