siRNA Details
| Gene Symbol: DCAKD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128631.2 | NP_001122103.1 | 5 | 1498 | ||
| NM_001288654.2 | NP_001275583.1 | 5 | 1634 | |||
| NM_001288655.1 | NP_001275584.1 | 5 | 1546 | |||
| NM_001321326.1 | NP_001308255.1 | 5 | 1444 | |||
| NM_024819.6 | NP_079095.3 | 5 | 1631 | |||
| XM_011525262.2 | XP_011523564.1 | 5 | 1663 | |||
| XM_017025105.1 | XP_016880594.1 | 6 | 1736 | |||
| XM_017025106.1 | XP_016880595.1 | 6 | 1678 | |||
| GenBank mRNA | BC006472.1 | - | 5 | 1510 | 2822306 | |
| BC006546.2 | - | 5 | 1510 | 2822306 | ||
| BC018132.2 | - | 4 | 1382 | 3903964 | ||
| BC033299.1 | - | 4 | 1521 | 4153431 | ||

DCAKD
Genomic Map