siRNA Details
| Gene Symbol: C16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320540.1 | NP_001307469.1 | 4 | 301 | ||
| NM_001320541.1 | NP_001307470.1 | 4 | 342 | |||
| NM_001320542.1 | NP_001307471.1 | 4 | 327 | |||
| NM_025187.4 | NP_079463.2 | 3 | 357 | |||
| XM_006721284.3 | XP_006721347.1 | 4 | 325 | |||
| XM_011523350.2 | XP_011521652.1 | 3 | 465 | |||
| XM_017023729.1 | XP_016879218.1 | 4 | 336 | |||
| XM_017023730.1 | XP_016879219.1 | 4 | 339 | |||
| XM_017023731.1 | XP_016879220.1 | 4 | 337 | |||
| GenBank mRNA | AF176088.1 | - | 4 | 257 | ||
| AK022138.1 | - | 2 | 126 | |||
| AL540304.3 | - | 4 | 236 | |||
| BC004556.2 | - | 3 | 337 | 3957147 | ||
| HY121038.1 | - | 3 | 358 | |||

C16orf70
Genomic Map