siRNA Details
| Gene Symbol: C16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320540.1 | NP_001307469.1 | 14 | 1147 | ||
| NM_001320541.1 | NP_001307470.1 | 14 | 1188 | |||
| NM_001320542.1 | NP_001307471.1 | 14 | 1173 | |||
| NM_001320543.1 | NP_001307472.1 | 13 | 1104 | |||
| NM_025187.4 | NP_079463.2 | 13 | 1203 | |||
| XM_006721284.3 | XP_006721347.1 | 13 | 1462 | |||
| XM_011523350.2 | XP_011521652.1 | 12 | 1602 | |||
| XM_017023729.1 | XP_016879218.1 | 13 | 1473 | |||
| GenBank mRNA | AF176088.1 | - | 14 | 1103 | ||
| BC004556.2 | - | 13 | 1183 | 3957147 | ||

C16orf70
Genomic Map