siRNA Details
| Gene Symbol: PLEKHO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195059.1 | NP_001181988.1 | 2, 3 | 271 | ||
| NM_025201.4 | NP_079477.2 | 3, 4 | 421 | |||
| GenBank mRNA | AF318373.1 | - | 19 | |||
| AF494534.1 | - | 3, 4 | 370 | |||
| AK096203.1 | - | 3, 4 | 389 | |||
| BC008744.2 | - | 2, 3 | 232 | 3346334 | ||

PLEKHO2
Genomic Map