siRNA Details
| Gene Symbol: AARSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001261434.1 | NP_001248363.1 | 6 | 701 | ||
| GenBank mRNA | AL515977.3 | - | 6 | 646 | ||
| Gene Symbol: PTGES3L-AARSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136042.2 | NP_001129514.2 | 11 | 1511 | ||
| NM_025267.3 | NP_079543.1 | 11 | 1127 | |||
| GenBank mRNA | AF039235.1 | - | 11 | 1216 | ||
| AK094084.1 | - | 11 | 1065 | |||
| AK295447.1 | - | 11 | 1511 | |||
| BC004172.2 | - | 11 | 1127 | 2823004 | ||
| BC019324.1 | - | 11 | 1127 | 2823004 | ||

AARSD1
Genomic Map