siRNA Details
| Gene Symbol: LIMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030576.3 | NP_085053.1 | 5 | 1426 | ||
| GenBank mRNA | AK092301.1 | - | 5 | 1392 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LIMD2| Gene Symbol: LIMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030576.3 | NP_085053.1 | 5 | 1426 | ||
| GenBank mRNA | AK092301.1 | - | 5 | 1392 | ||