siRNA Details
| Gene Symbol: GFOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030819.3 | NP_110446.3 | 3 | 2031 | ||
| NR_027398.1 | 2 | 1685 | ||||
| XM_006721288.3 | XP_006721351.1 | 3 | 1774 | |||
| XM_017023738.1 | XP_016879227.1 | 3 | 1981 | |||
| GenBank mRNA | AK074382.1 | - | 4 | 1984 | ||
| AK091606.1 | - | 1 | 2908 | |||
| AK225997.1 | - | 3 | 2001 | |||
| AL832904.1 | - | 1 | 1470 | |||
| AY358169.1 | - | 2 | 1741 | |||
| BC000757.2 | - | 2 | 1638 | 2820364 | ||
| BC002943.1 | - | 2 | 1658 | |||
| BC107585.1 | - | 3 | 1981 | 6463662 | ||

GFOD2
Genomic Map