siRNA Details
| Gene Symbol: YPEL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145524.1 | NP_001138996.1 | 5 | 798 | ||
| NM_031477.4 | NP_113665.3 | 4 | 1444 | |||
| GenBank mRNA | AB098737.1 | - | 5 | 798 | ||
| AF305622.1 | - | 5 | 801 | |||
| BC005009.1 | - | 5 | 754 | 3639517 | ||
| BC050664.1 | - | 4 | 1443 | 5748830 | ||

YPEL3
Genomic Map