siRNA Details
| Gene Symbol: LONP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300948.1 | NP_001287877.1 | 3 | 725 | ||
| NM_031490.3 | NP_113678.2 | 4 | 857 | |||
| XM_017023755.1 | XP_016879244.1 | 4 | 724 | |||
| XM_017023756.1 | XP_016879245.1 | 3 | 590 | |||
| GenBank mRNA | AJ548761.1 | - | 4 | 724 | ||
| AK027544.1 | - | 4 | 688 | |||
| AK074775.1 | - | 4 | 722 | |||
| AK127125.1 | - | 4 | 488 | |||
| AK294779.1 | - | 3 | 537 | |||
| AL834201.1 | - | 3 | 224 | |||
| BC093910.1 | - | 4 | 691 | 7939755 | ||
| BC093912.1 | - | 4 | 691 | 7939757 | ||
| BC143246.1 | - | 3 | 559 | 9051747 | ||

LONP2
Genomic Map