siRNA Details
| Gene Symbol: QRICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032134.2 | NP_115510.1 | 2 | 257 | ||
| NR_130649.1 | 2 | 257 | ||||
| XM_005257728.3 | XP_005257785.1 | 2 | 1267 | |||
| XM_017025206.1 | XP_016880695.1 | 2 | 1267 | |||
| XM_017025208.1 | XP_016880697.1 | 1 | 345 | |||
| GenBank mRNA | AL136774.1 | - | 2 | 257 | ||
| BC131559.1 | - | 2 | 142 | |||

QRICH2
Genomic Map