siRNA Details
| Gene Symbol: APOPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302652.1 | NP_001289581.1 | 5 | 1186 | ||
| NM_001302653.1 | NP_001289582.1 | 6 | 1276 | |||
| NM_032374.4 | NP_115750.2 | 5 | 1188 | |||
| NR_126431.1 | 6 | 1353 | ||||
| NR_126432.1 | 4 | 1124 | ||||
| GenBank mRNA | BC007412.2 | - | 5 | 1151 | 2989951 | |
| BP317962.2 | - | 4 | 462 | |||

APOPT1
Genomic Map