siRNA Details
| Gene Symbol: C15orf41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321756.1 | NP_001308685.1 | 10 | 1196 | ||
| NM_001321758.1 | NP_001308687.1 | 9 | 1131 | |||
| NM_001321759.1 | NP_001308688.1 | 11 | 1242 | |||
| NM_001321760.1 | NP_001308689.1 | 10 | 1136 | |||
| NM_001321761.1 | NP_001308690.1 | 11 | 1260 | |||
| NM_032499.5 | NP_115888.1 | 12 | 988 | |||
| XM_017022676.1 | XP_016878165.1 | 8 | 1036 | |||
| GenBank mRNA | AF114263.1 | - | 12 | 963 | ||

C15orf41
Genomic Map