siRNA Details
| Gene Symbol: CRACR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144958.1 | NP_001138430.1 | 10 | 1501 | ||
| NM_032680.3 | NP_116069.1 | 10 | 1501 | |||
| XM_006719021.3 | XP_006719084.1 | 10 | 1501 | |||
| XM_011521034.2 | XP_011519336.1 | 9 | 1248 | |||
| XM_011521036.2 | XP_011519338.1 | 8 | 1008 | |||
| XM_011521037.2 | XP_011519339.1 | 4 | 346 | |||
| XM_011521038.2 | XP_011519340.1 | 4 | 343 | |||
| GenBank mRNA | AK091124.1 | - | 4 | 337 | ||
| AK304017.1 | - | 10 | 1403 | |||
| BC004524.2 | - | 10 | 1398 | |||
| BC150643.1 | - | 9 | 1132 | 9057013 | ||

CRACR2A
Genomic Map