siRNA Details
| Gene Symbol: FAM104A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098832.1 | NP_001092302.1 | 4 | 864 | ||
| NM_001289410.1 | NP_001276339.1 | 2 | 701 | |||
| NM_001289411.1 | NP_001276340.1 | 3 | 719 | |||
| NM_001289412.1 | NP_001276341.1 | 4 | 637 | |||
| NM_032837.2 | NP_116226.2 | 3 | 801 | |||
| GenBank mRNA | AK027681.1 | - | 3 | 686 | ||
| AK074755.1 | - | 3 | 687 | |||
| AK304556.1 | - | 3 | 774 | |||
| BC011054.1 | - | 3 | 526 | 4177665 | ||
| BC025238.1 | - | 3 | 564 | 4905931 | ||
| BX537858.1 | - | 4 | 768 | |||
| BX648968.1 | - | 5 | 780 | |||
| U92989.1 | - | 4 | 608 | |||

FAM104A
Genomic Map