siRNA Details
| Gene Symbol: TNS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032865.5 | NP_116254.4 | 13 | 3864 | ||
| XM_005257744.1 | XP_005257801.1 | 13 | 3940 | |||
| XM_017025236.1 | XP_016880725.1 | 12 | 3865 | |||
| GenBank mRNA | AF370421.1 | - | 13 | 3851 | ||
| AF417488.1 | - | 13 | 3817 | |||
| AK027856.1 | - | 11 | 3205 | |||
| BC013373.2 | - | 4 | 1970 | |||
| BC018706.1 | - | 12 | 3298 | 5013235 | ||

TNS4
Genomic Map