siRNA Details
| Gene Symbol: FBXL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184906.1 | NP_001171835.1 | 13, 14 | 1371 | ||
| NM_032875.2 | NP_116264.2 | 14, 15 | 1467 | |||
| XM_005257746.3 | XP_005257803.2 | 14, 15 | 1658 | |||
| XM_005257747.3 | XP_005257804.2 | 13, 14 | 1439 | |||
| GenBank mRNA | AK123048.1 | - | 14, 15 | 1468 | ||
| AK291844.1 | - | 14, 15 | 1478 | |||
| AK312612.1 | - | 14, 15 | 1448 | |||
| BC007557.1 | - | 14, 15 | 1434 | 2987858 | ||
| DQ223959.1 | - | 13, 14 | 1338 | |||

FBXL20
Genomic Map