siRNA Details
| Gene Symbol: MFSD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170790.1 | NP_001164261.1 | 2 | 1561 | ||
| NM_032889.4 | NP_116278.3 | 2 | 1288 | |||
| XM_005269197.1 | XP_005269254.1 | 3 | 1816 | |||
| XM_005269198.4 | XP_005269255.1 | 2 | 1238 | |||
| GenBank mRNA | AK074684.1 | - | 2 | 1285 | ||
| AK225998.1 | - | 2 | 1173 | |||
| AY358604.1 | - | 2 | 1217 | |||
| BC007703.2 | - | 1 | 1104 | 3948896 | ||
| BC067795.1 | - | 2 | 1216 | 5299341 | ||
| BX640649.1 | - | 2 | 1182 | |||

MFSD5
Genomic Map