siRNA Details
| Gene Symbol: FRMD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286490.1 | NP_001273419.1 | 15 | 1406 | ||
| NM_001286491.1 | NP_001273420.1 | 15 | 1790 | |||
| NM_001322949.1 | NP_001309878.1 | 14 | 1471 | |||
| NM_001322950.1 | NP_001309879.1 | 14 | 1471 | |||
| NM_001322951.1 | NP_001309880.1 | 13 | 1366 | |||
| NM_032892.4 | NP_116281.2 | 14 | 1471 | |||
| NR_104455.1 | 15 | 1787 | ||||
| XM_005254730.3 | XP_005254787.1 | 14 | 1376 | |||
| GenBank mRNA | AK090572.1 | - | 14 | 1403 | ||
| AK123018.1 | - | 15 | 1406 | |||
| AK290013.1 | - | 3 | 265 | |||
| BC043486.2 | - | 15 | 1537 | |||
| BC053647.1 | - | 14 | 1408 | 5578205 | ||

FRMD5
Genomic Map