siRNA Details
| Gene Symbol: TMEM241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318834.1 | NP_001305763.1 | 16 | 1928 | ||
| NM_032933.5 | NP_116322.3 | 15 | 1813 | |||
| NR_134875.1 | 14 | 1764 | ||||
| NR_134876.1 | 11 | 1684 | ||||
| NR_134877.1 | 14 | 1767 | ||||
| XM_011526233.2 | XP_011524535.1 | 16 | 1905 | |||
| XM_017026045.1 | XP_016881534.1 | 14 | 1727 | |||
| XM_017026049.1 | XP_016881538.1 | 15 | 1809 | |||
| XM_017026050.1 | XP_016881539.1 | 15 | 1831 | |||
| XM_017026051.1 | XP_016881540.1 | 15 | 1733 | |||
| GenBank mRNA | AK123623.1 | - | 11 | 1674 | ||
| AK126247.1 | - | 14 | 1756 | |||
| BC006280.2 | - | 3 | 1034 | |||
| BC054507.1 | - | 9 | 1310 | |||
| BC082984.1 | - | 3 | 1161 | |||

TMEM241
Genomic Map