siRNA Details
| Gene Symbol: ZNF764 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172679.1 | NP_001166150.1 | 3 | 2544 | ||
| NM_033410.3 | NP_219363.2 | 3 | 2547 | |||
| GenBank mRNA | BC000016.2 | - | 1 | 1780 | ||
| BC008821.2 | - | 3 | 2489 | 4130947 | ||

ZNF764
Genomic Map