siRNA Details
Gene Symbol: IFT43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102564.1 | NP_001096034.1 | 9 | 736 | ||
NM_052873.2 | NP_443105.2 | 8 | 751 | |||
GenBank mRNA | AK056735.1 | - | 9 | 1047 | ||
BC010436.1 | - | 8 | 722 | 3608220 |
