siRNA Details
| Gene Symbol: IFT43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052873.2 | NP_443105.2 | 4 | 294 | ||
| GenBank mRNA | AK056735.1 | - | 6 | 347 | ||
| BC010436.1 | - | 4 | 265 | 3608220 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IFT43| Gene Symbol: IFT43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052873.2 | NP_443105.2 | 4 | 294 | ||
| GenBank mRNA | AK056735.1 | - | 6 | 347 | ||
| BC010436.1 | - | 4 | 265 | 3608220 | ||