siRNA Details
| Gene Symbol: WIPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133264.4 | NP_573571.1 | 8 | 1955 | ||
| XM_005257083.2 | XP_005257140.1 | 8 | 1957 | |||
| XM_005257084.2 | XP_005257141.1 | 8 | 1915 | |||
| XM_011524412.1 | XP_011522714.1 | 7 | 1856 | |||
| XM_011524413.1 | XP_011522715.1 | 7 | 1837 | |||
| GenBank mRNA | AF370396.1 | - | 8 | 1880 | ||
| AK026913.1 | - | 6 | 1403 | |||
| AK055057.1 | - | 6 | 1172 | |||
| AK290301.1 | - | 8 | 1951 | |||
| AK315836.1 | - | 8 | 1935 | |||
| AL834239.1 | - | 7 | 1824 | |||
| BC025965.1 | - | 8 | 1911 | |||
| BC065551.1 | - | 8 | 1892 | 6044222 | ||
| U90911.1 | - | 1 | 378 | |||

WIPF2
Genomic Map