siRNA Details
| Gene Symbol: MARCH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138396.5 | NP_612405.2 | 4 | 1495 | ||
| GenBank mRNA | AK093897.1 | - | 2 | 410 | ||
| BC009489.2 | - | 2 | 975 | 3936171 | ||
| BC036455.2 | - | 4 | 1476 | 4811410 | ||
| BC050397.1 | - | 4 | 1495 | |||

MARCH9
Genomic Map