siRNA Details
| Gene Symbol: ZNF689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138447.2 | NP_612456.1 | 3 | 2413 | ||
| NR_073480.1 | 3 | 2048 | ||||
| NR_073481.1 | 3 | 2051 | ||||
| NR_073482.1 | 3 | 1931 | ||||
| NR_104166.1 | 3 | 1950 | ||||
| GenBank mRNA | AB065282.1 | - | 3 | 2398 | ||
| AF086510.1 | - | 1 | 350 | |||
| AK074896.1 | - | 3 | 2136 | |||
| AL834299.1 | - | 1 | 1219 | |||
| BC014000.2 | - | 3 | 2388 | 3936339 | ||

ZNF689
Genomic Map