siRNA Details
| Gene Symbol: IQCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138451.2 | NP_612460.1 | 3 | 1313 | ||
| XM_005253832.4 | XP_005253889.1 | 5 | 1862 | |||
| XM_005253833.4 | XP_005253890.1 | 5 | 2110 | |||
| XM_005253834.4 | XP_005253891.1 | 5 | 1322 | |||
| XM_011537862.2 | XP_011536164.1 | 5 | 2742 | |||
| XM_011537863.2 | XP_011536165.1 | 6 | 1988 | |||
| XM_011537864.1 | XP_011536166.1 | 4 | 1385 | |||
| XM_011537865.2 | XP_011536167.1 | 4 | 1089 | |||
| XM_011537866.2 | XP_011536168.1 | 3 | 806 | |||
| XM_017018771.1 | XP_016874260.1 | 3 | 1020 | |||
| GenBank mRNA | BC013151.1 | - | 3 | 1294 | 4385421 | |

IQCD
Genomic Map