siRNA Details
| Gene Symbol: PPTC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139283.1 | NP_644812.1 | 6 | 1156 | ||
| GenBank mRNA | AF385435.1 | - | 6 | 1156 | ||
| AK124744.1 | - | 7 | 1000 | |||
| BC111551.1 | - | 7 | 958 | 40009728 | ||
| CR749216.1 | - | 3 | 278 | |||

PPTC7
Genomic Map