siRNA Details
| Gene Symbol: LIMK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204426.1 | NP_001191355.1 | 9 | 1215 | ||
| NM_002314.3 | NP_002305.1 | 10 | 1437 | |||
| GenBank mRNA | AB209557.1 | - | 7 | 909 | ||
| AF134379.1 | - | 10 | 1162 | |||
| AK026544.1 | - | 4 | 473 | |||
| AK290162.1 | - | 11 | 1362 | |||
| AK300382.1 | - | 9 | 1154 | |||
| D26309.1 | - | 11 | 1318 | |||

LIMK1
Genomic Map