siRNA Details
| Gene Symbol: C16orf78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144602.3 | NP_653203.1 | 2, 3 | 407 | ||
| GenBank mRNA | BC021181.2 | - | 2, 3 | 318 | 5268227 | |
| BC021718.2 | - | 2, 3 | 318 | 5265370 | ||
| HY001804.1 | - | 2, 3 | 407 | |||

C16orf78
Genomic Map