siRNA Details
Gene Symbol: C16orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144602.3 | NP_653203.1 | 4 | 733 | ||
GenBank mRNA | BC021181.2 | - | 4 | 644 | 5268227 | |
BC021718.2 | - | 4 | 644 | 5265370 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C16orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144602.3 | NP_653203.1 | 4 | 733 | ||
GenBank mRNA | BC021181.2 | - | 4 | 644 | 5268227 | |
BC021718.2 | - | 4 | 644 | 5265370 |