siRNA Details
| Gene Symbol: SEPT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144605.4 | NP_653206.2 | 5 | 674 | ||
| XM_006720846.2 | XP_006720909.1 | 5 | 810 | |||
| XM_011522379.2 | XP_011520681.1 | 4 | 323 | |||
| XM_017022938.1 | XP_016878427.1 | 4 | 788 | |||
| GenBank mRNA | AK058139.1 | - | 5 | 500 | ||
| AK098718.1 | - | 5 | 501 | |||
| BC024017.1 | - | 5 | 474 | |||
| BC035619.1 | - | 5 | 480 | 5172555 | ||
| DQ517531.1 | - | 5 | 551 | |||
| EF620906.1 | - | 5 | 674 | |||

SEPT12
Genomic Map