siRNA Details
| Gene Symbol: CYB5D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144607.4 | NP_653208.2 | 4 | 1585 | ||
| XM_005256440.3 | XP_005256497.1 | 4 | 1296 | |||
| GenBank mRNA | AK022811.1 | - | 1 | 206 | ||
| AK057061.1 | - | 4 | 1517 | |||
| AK125598.1 | - | 1 | 1179 | |||
| AK289520.1 | - | 4 | 1268 | |||
| BC060779.1 | - | 4 | 1337 | 30349502 | ||

CYB5D1
Genomic Map