siRNA Details
| Gene Symbol: TEPSIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144679.2 | NP_653280.1 | 12 | 1719 | ||
| XM_005257066.2 | XP_005257123.1 | 14 | 2517 | |||
| XM_005257067.2 | XP_005257124.1 | 13 | 1923 | |||
| XM_006721709.3 | XP_006721772.1 | 13 | 1930 | |||
| XM_006721712.2 | XP_006721775.1 | 13 | 2361 | |||
| XM_011524355.1 | XP_011522657.1 | 14 | 2523 | |||
| XM_011524356.1 | XP_011522658.1 | 13 | 2319 | |||
| XM_011524357.1 | XP_011522659.1 | 13 | 2551 | |||
| XM_011524358.2 | XP_011522660.1 | 14 | 2452 | |||
| XM_017024202.1 | XP_016879691.1 | 14 | 2764 | |||
| XM_017024203.1 | XP_016879692.1 | 15 | 2411 | |||
| XM_017024204.1 | XP_016879693.1 | 14 | 2964 | |||
| GenBank mRNA | AK056090.1 | - | 12 | 1683 | ||
| AK127221.1 | - | 11 | 3076 | |||
| AK128728.1 | - | 14 | 3236 | |||
| BC064483.1 | - | 12 | 1680 | 5430837 | ||

TEPSIN
Genomic Map