siRNA Details
| Gene Symbol: CFAP53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145020.4 | NP_659457.2 | 4 | 612 | ||
| GenBank mRNA | AK057305.1 | - | 3 | 333 | ||
| AK302114.1 | - | 4 | 612 | |||
| BC030606.2 | - | 4 | 585 | 4831081 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CFAP53| Gene Symbol: CFAP53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145020.4 | NP_659457.2 | 4 | 612 | ||
| GenBank mRNA | AK057305.1 | - | 3 | 333 | ||
| AK302114.1 | - | 4 | 612 | |||
| BC030606.2 | - | 4 | 585 | 4831081 | ||