siRNA Details
| Gene Symbol: SPACA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145248.4 | NP_660291.2 | 3 | 253 | ||
| XM_011537466.1 | XP_011535768.1 | 3 | 278 | |||
| XM_011537467.2 | XP_011535769.1 | 2 | 221 | |||
| XM_011537468.2 | XP_011535770.1 | 4 | 339 | |||
| XM_011537471.2 | XP_011535773.1 | 4 | 409 | |||
| GenBank mRNA | AK130866.1 | - | 3 | 253 | ||
| BC016750.2 | - | 3 | 235 | 4837062 | ||

SPACA7
Genomic Map