siRNA Details
Gene Symbol: BEST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139443.1 | NP_001132915.1 | 7 | 1300 | ||
NM_001300787.1 | NP_001287716.1 | 7 | 1300 | |||
NM_004183.3 | NP_004174.1 | 8 | 1488 | |||
NR_134580.1 | 8 | 1691 | ||||
XM_005274210.3 | XP_005274267.1 | 8 | 1598 | |||
XM_005274215.3 | XP_005274272.1 | 6 | 1253 | |||
XM_005274216.3 | XP_005274273.1 | 7 | 1613 | |||
XM_011545229.2 | XP_011543531.1 | 8 | 1038 | |||
XM_011545230.2 | XP_011543532.1 | 7 | 841 | |||
XM_011545233.2 | XP_011543535.1 | 2 | 735 | |||
XM_017018228.1 | XP_016873717.1 | 6 | 1253 | |||
XM_017018229.1 | XP_016873718.1 | 8 | 1801 | |||
XM_017018230.1 | XP_016873719.1 | 6 | 1456 | |||
GenBank mRNA | AB209451.1 | - | 5 | 2529 | ||
AF057169.1 | - | 8 | 1012 | |||
AF057170.1 | - | 8 | 1215 | |||
AF073501.1 | - | 8 | 996 | |||
AK289681.1 | - | 8 | 1219 | |||
AK293701.1 | - | 7 | 954 | |||
AK295450.1 | - | 7 | 1034 | |||
AK295998.1 | - | 7 | 834 | |||
AK301392.1 | - | 6 | 817 | |||
AY515704.1 | - | 7 | 908 | |||
BC041664.1 | - | 7 | 814 | 5194649 | ||
JQ954696.1 | - | 7 | 974 | |||
KU178391.1 | - | 6 | 728 | |||
KU178392.1 | - | 5 | 575 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
