siRNA Details
| Gene Symbol: WDR90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145294.4 | NP_660337.3 | 41 | 5420 | ||
| XM_017023023.1 | XP_016878512.1 | 42 | 5690 | |||
| XM_017023024.1 | XP_016878513.1 | 24 | 3303 | |||
| GenBank mRNA | AK091062.1 | - | 14 | 2610 | ||
| AK092719.1 | - | 12 | 2136 | |||
| AK093609.1 | - | 6 | 1820 | |||
| AK131510.1 | - | 24 | 3726 | |||
| BC065836.1 | - | 6 | 1332 | |||

WDR90
Genomic Map