siRNA Details
| Gene Symbol: RPUSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286407.1 | NP_001273336.1 | 3 | 829 | ||
| NM_152260.2 | NP_689473.1 | 3 | 1012 | |||
| GenBank mRNA | AK055971.1 | - | 3 | 967 | ||
| AK293145.1 | - | 3 | 790 | |||
| AK301383.1 | - | 4 | 985 | |||
| AK301706.1 | - | 4 | 904 | |||
| BC007697.1 | - | 3 | 994 | |||
| BC016967.1 | - | 3 | 973 | 4420027 | ||

RPUSD2
Genomic Map