siRNA Details
| Gene Symbol: RMI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152308.2 | NP_689521.1 | 2 | 1064 | ||
| NR_130754.1 | 2 | 841 | ||||
| GenBank mRNA | AK123764.1 | - | 2 | 1053 | ||
| BC013040.2 | - | 2 | 1032 | 3542171 | ||
| BC022427.1 | - | 2 | 1048 | 4252688 | ||
| BC031016.1 | - | 2 | 1052 | 4659915 | ||
| BC039361.1 | - | 5 | 1486 | 5270594 | ||

RMI2
Genomic Map