siRNA Details
| Gene Symbol: GLIPR1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270396.1 | NP_001257325.1 | 1 | 202 | ||
| NM_152436.2 | NP_689649.1 | 1 | 202 | |||
| NR_072995.1 | 1 | 202 | ||||
| XM_011537949.2 | XP_011536251.1 | 1 | 183 | |||
| XM_017018848.1 | XP_016874337.1 | 11 | ||||
| GenBank mRNA | AK093175.1 | - | 1 | 192 | ||
| BC016749.1 | - | 1 | 209 | |||
| BC029557.1 | - | 1 | 163 | 5262538 | ||
| BX640916.1 | - | 1 | 171 | |||

GLIPR1L2
Genomic Map