siRNA Details
| Gene Symbol: C12orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300940.1 | NP_001287869.1 | 3 | 944 | ||
| NM_001300941.1 | NP_001287870.1 | 3 | 785 | |||
| NM_152440.4 | NP_689653.3 | 3 | 944 | |||
| GenBank mRNA | AK057111.1 | - | 3 | 911 | ||
| AK129933.1 | - | 3 | 785 | |||
| BC036246.1 | - | 3 | 913 | |||
| BC071634.1 | - | 3 | 901 | 5182140 | ||

C12orf66
Genomic Map