siRNA Details
| Gene Symbol: ZSCAN29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152455.3 | NP_689668.3 | 2, 3 | 663 | ||
| XM_006720401.3 | XP_006720464.1 | 3, 4 | 1423 | |||
| XM_011521266.2 | XP_011519568.1 | 3, 4 | 1103 | |||
| XM_017021949.1 | XP_016877438.1 | 2, 3 | 1635 | |||
| GenBank mRNA | AF525399.1 | - | 2, 3 | 513 | ||
| AK093186.1 | - | 1, 2 | 209 | |||
| AK122788.1 | - | 2, 3 | 651 | |||
| BC062551.1 | - | 2, 3 | 279 | |||
| BC109270.1 | - | 2, 3 | 513 | 40026738 | ||

ZSCAN29
Genomic Map